missing translation for 'onlineSavingsMsg'
Learn More
1,5-Diphenylcarbazide, 97+%, Thermo Scientific Chemicals Product Code.: 11407986

1,5-Diphenylcarbazide, 97+%, Thermo Scientific Chemicals

Product Code. 11407986
500 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11407986

Brand: Thermo Scientific Alfa Aesar A14060.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1,5-Diphenylcarbazide is used in the colorimteric determination of chromium, osmium as well as in the detection of cadmium, mercury, magnesium, emetine and aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,5-Diphenylcarbazide is used in the colorimteric determination of chromium, osmium as well as in the detection of cadmium, mercury, magnesium, emetine and aldehydes.

Solubility
Soluble in glacial acetic acid, alcohol and acetone. Slightly soluble in water.

Notes
Air and light sensitive. Keep the container tightly closed in a dry and well-ventilated place.Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 140-22-7
Molecular Formula C13H14N4O
Molecular Weight (g/mol) 242.282
MDL Number MFCD00003013
InChI Key KSPIHGBHKVISFI-UHFFFAOYSA-N
Synonym 1,5-diphenylcarbazide, diphenylcarbazide, 1,5-diphenylcarbohydrazide, carbonic dihydrazide, 2,2'-diphenyl, 1,5-diphenylcarbonohydrazide, 1,5-diphenylcabohydrazide, sym-diphenylcarbazide, n,n'-diphenylcarbazide, 2,2'-diphenylcarbazide, carbohydrazide, 1,5-diphenylShow MoreShow Less
PubChem CID 8789
ChEBI CHEBI:4641
IUPAC Name 1,3-dianilinourea
SMILES C1=CC=C(C=C1)NNC(=O)NNC2=CC=CC=C2

Specifications

Melting Point 170°C to 176°C
Quantity 500 g
Beilstein 752039
Sensitivity Air and light sensitive
Merck Index 14,3321
Solubility Information Soluble in glacial acetic acid,alcohol and acetone. Slightly soluble in water.
Formula Weight 242.28
Percent Purity ≥97%
Chemical Name or Material 1,5-Diphenylcarbazide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10249513 Certificate TypeCertificate of Analysis Date02/02/2024 Catalog NumberA14060.22,A14060.14,A14060.36
Lot NumberU08I043 Certificate TypeCertificate of Analysis Date27/10/2022 Catalog NumberA14060.22,A14060.14,A14060.36
Lot Number10242609 Certificate TypeCertificate of Analysis Date06/08/2022 Catalog NumberA14060.22,A14060.14,A14060.36
Lot NumberQ16H044 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA14060.22,A14060.14,A14060.36
Lot NumberW24H057 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA14060.22,A14060.14,A14060.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,5-Diphenylcarbazide, 97+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.