missing translation for 'onlineSavingsMsg'
Learn More
1-(3-Chlorophenyl)-2-(isopropylamino)ethanol, Thermo Scientific™ Product Code.: 11414670

1-(3-Chlorophenyl)-2-(isopropylamino)ethanol, Thermo Scientific™

Product Code. 11414670
250 mg, 250mg
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11414670

Brand: Thermo Scientific Alfa Aesar H50739.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23299-18-5
Molecular Formula C11H16ClNO
Molecular Weight (g/mol) 213.705
MDL Number MFCD08060964
InChI Key WPRYXHDMUFXECU-UHFFFAOYSA-N
Synonym 1-3-chlorophenyl-2-isopropylamino ethanol, 1-3-chloro-phenyl-2-isopropylamino-ethanol, 1-3-chlorophenyl-2-methylethyl amino ethan-1-ol, 1-3-chlorophenyl-2-propan-2-yl amino ethan-1-ol, benzenemethanol,3-chloro-a-1-methylethyl amino methyl
PubChem CID 19913640
IUPAC Name 1-(3-chlorophenyl)-2-(propan-2-ylamino)ethanol
SMILES CC(C)NCC(C1=CC(=CC=C1)Cl)O

Specifications

Quantity 250 mg
Formula Weight 213.71
Chemical Name or Material 1-(3-Chlorophenyl)-2-(isopropylamino)ethanol

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Product Code
Lot NumberK30Q013 Certificate TypeCertificate of Analysis Date08/12/2021 Product Code
Lot NumberE24Z017 Certificate TypeCertificate of Analysis Date06/12/2021 Product Code
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-(3-Chlorophenyl)-2-(isopropylamino)ethanol, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.