missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Methoxyphenyl)piperazine hydrochloride, 97%, Thermo Scientific™

Product Code. 11484707
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1-(2-Methoxyphenyl)piperazine hydrochloride is used as reference material for forensic laboratories. It is used in chemical synthesis studies.

Solubility
Soluble in DMSO and Methanol.

Notes
Hygroscopic. Store at -20°C. Store away from oxidizing agents. Store in a cool, dry condition in a well sealed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5464-78-8
Molecular Formula C11H17ClN2O
Molecular Weight (g/mol) 228.72
MDL Number MFCD00012762
InChI Key DDMVHGULHRJOEC-UHFFFAOYSA-N
Synonym 1-2-methoxyphenyl piperazine hydrochloride, ompp, unii-3xq74ihw73, 1-2-methoxyphenyl piperazine hcl, 1-2-methoxyphenyl-pepipazine hcl, n-2-methoxyphenyl piperazine hydrochloride, 1 2-methoxyphenyl piperazine hydrochloride, 1-2-methoxylphenyl piperazine hydrochloride, dsstox_cid_30037, dsstox_rid_81139
PubChem CID 9899402
IUPAC Name 1-(2-methoxyphenyl)piperazine;hydrochloride
SMILES COC1=CC=CC=C1N2CCNCC2.Cl

Specifications

Melting Point 213°C to 216°C
Quantity 100 g
UN Number UN2811
Beilstein 4276925
Sensitivity Hygroscopic
Solubility Information Soluble in DMSO and Methanol.
Formula Weight 228.72
Percent Purity 97%
Chemical Name or Material 1-(2-Methoxyphenyl)piperazine hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.