Learn More
1,2-Dibromoethane, 99%, Thermo Scientific Chemicals Product Code.: 15473057
1,2-Dibromoethane, 99%, Thermo Scientific Chemicals
Product Code. 15473057
1000 g
Click to view available options
Quantity:
250 g
1000 g
Product Code. 15473057
Brand: Thermo Scientific Alfa Aesar A12766.0B
Request Bulk or Custom Format
1,2-dibromoethane is widely used as a bromine source in organic synthesis. It is utilized as a fumigant for treatment of logs for termites and beetles, for control of moths in beehives. It finds the application to make vinyl bromide, a precursor to some fire retardants. It is used as an intermediate in the preparation of dyes and pharmaceuticals. It is also used as a lead scavenger in antiknock mixtures added to gasolines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,2-dibromoethane is widely used as a bromine source in organic synthesis. It is utilized as a fumigant for treatment of logs for termites and beetles, for control of moths in beehives. It finds the application to make vinyl bromide, a precursor to some fire retardants. It is used as an intermediate in the preparation of dyes and pharmaceuticals. It is also used as a lead scavenger in antiknock mixtures added to gasolines.
Solubility
Miscible with alcohol, acetone, benzene, diethyl ether and ethyl acetate. Slightly miscible with water.
Notes
Special handling precautions required. View MSDS prior to purchase. MSDS are available online at www.alfa.com.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 106-93-4
Molecular Formula C2H4Br2
Molecular Weight (g/mol) 187.862
MDL Number MFCD00000233
InChI Key PAAZPARNPHGIKF-UHFFFAOYSA-N
Synonym ethylene dibromide, ethylene bromide, sym-dibromoethane, ethane, 1,2-dibromo, alpha,beta-dibromoethane, bromuro di etile, 1,2-dibromaethan, 1,2-dibroomethaan, 1,2-ethylene dibromide, aadibroomShow MoreShow Less
PubChem CID 7839
ChEBI CHEBI:28534
IUPAC Name 1,2-dibromoethane
SMILES C(CBr)Br
Specifications
Melting Point 9°C to 10°C
Density 2.179
Boiling Point 132°C
Flash Point None
Linear Formula BrCH2CH2Br
Refractive Index 1.5385
Quantity 1000 g
UN Number UN1605
Sensitivity Light sensitive
Merck Index 14,3796
Solubility Information Miscible with alcohol,acetone,benzene,diethyl ether and ethyl acetate. Slightly miscible with water.
Formula Weight 187.87
Percent Purity 99%
Chemical Name or Material 1,2-Dibromoethane
Show More Show Less
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0461610 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog NumberA12766.0B,A12766.0P,A12766.30
Lot NumberU20K032 Certificate TypeCertificate of Analysis Date11/09/2024 Catalog NumberA12766.0B,A12766.0P,A12766.30
Lot NumberP04K042 Certificate TypeCertificate of Analysis Date19/03/2024 Catalog NumberA12766.0B,A12766.0P,A12766.30
Lot Number10215822 Certificate TypeCertificate of Analysis Date22/12/2023 Catalog NumberA12766.0B,A12766.0P,A12766.30,A12766.0I
Lot NumberY08J033 Certificate TypeCertificate of Analysis Date15/11/2023 Catalog NumberA12766.0B,A12766.0P,A12766.30,A12766.0I
Product Title
1,2-Dibromoethane, 99%, Thermo Scientific Chemicals >