missing translation for 'onlineSavingsMsg'
Learn More
1-(1-Naphthylmethyl)piperazine, 97% Product Code.: 15421628

1-(1-Naphthylmethyl)piperazine, 97%

Product Code. 15421628
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 15421628

Brand: Thermo Scientific Alfa Aesar H55398.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 40675-81-8 | C15H18N2 | 226.323 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40675-81-8
Molecular Formula C15H18N2
Molecular Weight (g/mol) 226.323
MDL Number MFCD01314185
InChI Key HGYDREHWXXUUIS-UHFFFAOYSA-N
Synonym 1-1-naphthylmethyl piperazine, 1-naphthalen-1-ylmethyl piperazine, 1-1-naphthylmethyl-piperazine, 1-naphthalen-1-ylmethyl-piperazine, 1-naphthalen-1-yl methyl piperazine, naphthylmethyl piperazine, 1-naphthylmethyl piperazine, 1-piperazin-1-yl naphthalene, 1-naphth-1-ylmethyl piperazine, 1-1-naphthalenylmethyl piperazineShow MoreShow Less
PubChem CID 701891
IUPAC Name 1-(naphthalen-1-ylmethyl)piperazine
SMILES C1CN(CCN1)CC2=CC=CC3=CC=CC=C32

Specifications

Melting Point 63°C to 67°C
Color White to Brown
Quantity 5 g
UN Number UN3077
Formula Weight 226.32
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1-(1-Naphthylmethyl)piperazine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberQ16L039 Certificate TypeCertificate of Analysis Date17/05/2025 Catalog NumberH55398.06,H55398.14,H55398.03
Lot NumberP26L024 Certificate TypeCertificate of Analysis Date28/03/2025 Catalog NumberH55398.06,H55398.14,H55398.03
Lot NumberU27K051 Certificate TypeCertificate of Analysis Date11/09/2024 Catalog NumberH55398.06,H55398.03,H55398.14
Lot NumberM06I019 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberH55398.06,H55398.03,H55398.14
Lot Number61600495 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH55398.06,H55398.03,H55398.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-(1-Naphthylmethyl)piperazine, 97% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.