missing translation for 'onlineSavingsMsg'
Learn More
1,1'-Diethyl-4,4'-carbocyanine Iodide Product Code.: 11421556

1,1'-Diethyl-4,4'-carbocyanine Iodide

Product Code. 11421556 Shop All MP Biomedicals Products
100 mg, 100mg
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 11421556

Brand: MP Biomedicals 0520715680

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 4727-50-8
Molecular Formula C25H25IN2
Quantity 100 mg
Synonym cryptocyanin, cryptocyanine, kryptocyanin, kryptocyanine, cryptocyanine iodide, kryptocyanine iodide, 1,1'-diethylquino-4-carbocyanine iodide, cryptocyanine o.a. 2, 1,4'-carbocyanine iodide, 1,4'-quinocarbocyanine iodide
InChI Key CEJANLKHJMMNQB-UHFFFAOYSA-M
SMILES CCN1C=CC(=CC=CC2=CC=[N+](C3=CC=CC=C23)CC)C4=CC=CC=C41.[I-]
IUPAC Name (4E)-1-ethyl-4-[(Z)-3-(1-ethylquinolin-1-ium-4-yl)prop-2-enylidene]quinoline;iodide
Molecular Weight (g/mol) 480.393
PubChem CID 16219292
ChEBI CHEBI:51502
Formula Weight 480.39
Chemical Name or Material 1, 1«-Diethyl-4, 4«-Carbocyanine Iodide
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1,1'-Diethyl-4,4'-carbocyanine Iodide >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.