Learn More
1,1,2,2-Tetrachloroethane-d2 99 atom% D, Thermo Scientific Chemicals
CAS: 33685-54-0 | C2H2Cl4 | 169.85 g/mol
1142.81 NOK - 3339.06 NOK
Chemical Identifiers
CAS | 33685-54-0 |
---|---|
Molecular Formula | C2H2Cl4 |
Molecular Weight (g/mol) | 169.85 |
MDL Number | MFCD00037672 |
InChI Key | QPFMBZIOSGYJDE-QDNHWIQGSA-N |
Synonym | 1,1,2,2-tetrachloroethane-d2, 1,2-dideutero-1,1,2,2-tetrachloroethane, tetrachloro 2 h? ethane, 1,1,2,2-tetrachloroethane-d2 tce-d2, ethane-d2, 1,1,2,2-tetrachloro, ethane-1,2-d2, 1,1,2,2-tetrachloro, 1,1,2,2-tetrachloro-1,2-2h2 ethane, de87d, 1,1,2,2-tetrachloroethane-d2 >99.50 atom % d, 1,1,2,2-tetrachloroethane-d2, ≥99.5 atom % d |
PubChem CID | 118531 |
IUPAC Name | 1,1,2,2-tetrachloro-1,2-dideuterioethane |
SMILES | C(C(Cl)Cl)(Cl)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10705801
|
Thermo Scientific Acros
133580050 |
5 mL | Glass Bottle |
1142.81 NOK
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10225140
|
Thermo Scientific Acros
133580200 |
20 mL | Glass Bottle |
3339.06 NOK
20mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMRChemical Identifiers
33685-54-0 | |
169.85 | |
QPFMBZIOSGYJDE-QDNHWIQGSA-N | |
118531 | |
C(C(Cl)Cl)(Cl)Cl |
C2H2Cl4 | |
MFCD00037672 | |
1,1,2,2-tetrachloroethane-d2, 1,2-dideutero-1,1,2,2-tetrachloroethane, tetrachloro 2 h? ethane, 1,1,2,2-tetrachloroethane-d2 tce-d2, ethane-d2, 1,1,2,2-tetrachloro, ethane-1,2-d2, 1,1,2,2-tetrachloro, 1,1,2,2-tetrachloro-1,2-2h2 ethane, de87d, 1,1,2,2-tetrachloroethane-d2 >99.50 atom % d, 1,1,2,2-tetrachloroethane-d2, ≥99.5 atom % d | |
1,1,2,2-tetrachloro-1,2-dideuterioethane |
Specifications
Liquid | |
1.6100g/mL | |
1.4924 | |
146.0°C | |
(Cl2CD)2 | |
01, III, 164 | |
QPFMBZIOSGYJDE-QDNHWIQGSA-N | |
1,1,2,2-tetrachloro-1,2-dideuterioethane | |
118531 | |
NMR |
Colorless | |
98.5% min. | |
33685-54-0 | |
C2H2Cl4 | |
MFCD00037672 | |
1,1,2,2-tetrachloroethane-d2, 1,2-dideutero-1,1,2,2-tetrachloroethane, tetrachloro 2 h? ethane, 1,1,2,2-tetrachloroethane-d2 tce-d2, ethane-d2, 1,1,2,2-tetrachloro, ethane-1,2-d2, 1,1,2,2-tetrachloro, 1,1,2,2-tetrachloro-1,2-2h2 ethane, de87d, 1,1,2,2-tetrachloroethane-d2 >99.50 atom % d, 1,1,2,2-tetrachloroethane-d2, ≥99.5 atom % d | |
C(C(Cl)Cl)(Cl)Cl | |
169.85 | |
169.86 | |
1,1,2,2-Tetrachloroethane-d2, 99 atom % D |
Safety and Handling
GHS H Statement:
Fatal if inhaled.
Fatal in contact with skin.
Toxic to aquatic life with long lasting effects.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 251-634-1
missing translation for 'tsca' : TSCA
- 1,1,2,2-Tetrachloroethane-d2
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 1
- Acute toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H330-Fatal if inhaled.
- H411-Toxic to aquatic life with long lasting effects.
- H310-Fatal in contact with skin.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.