missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG Antibody (97924) [DyLight 488], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB110K
This item is not returnable.
View return policy
Description
IgG Monoclonal antibody specifically detects IgG in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
IgG | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, CyTOF | |
97924 | |
Western Blot, Flow Cytometry, CyTOF-ready | |
Immunoglobulin G, ImmunoglobulinG | |
Mouse myeloma cell line NS0-derived recombinant human IgG1 Fc region | |
0.1 mL | |
Adaptive Immunity, Epitope Tags, Immunology | |
3500 | |
Store at 4°C in the dark. | |
IgG1 |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 12
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
IgG Antibody (97924) [DyLight 488], Novus Biologicals™ > 0.1 mL; DyLight 488
Spot an opportunity for improvement?Share a Content Correction