missing translation for 'onlineSavingsMsg'
Learn More

HspA6 Antibody, CoraFluor™ 1, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals (Bio-Techne) NBP2-97536CL1

Product Code. 30144309

  • 7430.00 NOK / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

HspA6 Polyclonal antibody specifically detects HspA6 in Human samples. It is validated for Western Blot,Immunocytochemistry/Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HspA6
Polyclonal
PBS
Rabbit
Antigen and protein A Affinity-purified
RUO
3310
Store at 4°C in the dark. Do not freeze.
IgG
Western Blot, Immunocytochemistry/Immunofluorescence
CoraFluor 1
heat shock 70 kDa protein 6, Heat shock 70 kDa protein B', heat shock 70kD protein 6 (HSP70B'), heat shock 70kDa protein 6 (HSP70B'), HSP70B'
Produced in rabbits immunized with E. coli-derived Human HspA6 fragment.
0.1 mL
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
HspA6 Antibody, CoraFluor™ 1, Novus Biologicals™ > 0.1 mL; CoraFluor 1

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.