accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 99768-12-4

CH3OOBOHHO

CAS RN 99768-12-4

IUPAC Name: [4-(methoxycarbonyl)phenyl]boronic acid
Synonyms: 4-(methoxycarbonyl)phenylboronic acid
Molecular Weight (g/mol): 179.97
Molecular Formula: C8H9BO4
InChi Key: PQCXFUXRTRESBD-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=C(C=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.