accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 97753-82-7

HO(R)O(S)OHNBr(R)OH(S)HO(R)HO

CAS RN 97753-82-7

IUPAC Name: (2S,3R,4S,5R,6R)-2-[(5-bromo-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
Synonyms: (2S,3R,4S,5R,6R)-2-[(5-bromo-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Weight (g/mol): 374.19
Molecular Formula: C14H16BrNO6
InChi Key: LINMATFDVHBYOS-MBJXGIAVSA-N
SMILES: OC[C@H]1O[C@@H](OC2=CNC3=CC=C(Br)C=C23)[C@H](O)[C@@H](O)[C@H]1O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.