accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 95737-68-1

CH3OOON

CAS RN 95737-68-1

IUPAC Name: 2-{[1-(4-phenoxyphenoxy)propan-2-yl]oxy}pyridine
Synonyms: pyriproxyfen pyriproxyfen, (s)-isomer (r)-isomer pyriproxyfen (R)-isomer pyriproxyfen (S)-pyriproxyfen pyriproxyfen, (R)-isomer (s)-isomer pyriproxyfen (S)-isomer pyriproxyfen (R)-pyriproxyfen pyriproxyfen, (R)-
Molecular Weight (g/mol): 321.38
Molecular Formula: C20H19NO3
InChi Key: NHDHVHZZCFYRSB-UHFFFAOYNA-N
SMILES: CC(COC1=CC=C(OC2=CC=CC=C2)C=C1)OC1=CC=CC=N1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Concentration 
  • (2)
  • (1)
Element 
  • (2)
  • (1)
Solvent or Matrix 
  • (2)
  • (1)
Grade 
  • (1)
  • (1)

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.