accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 95-45-4

CH3NOHH3CNHO(E)(E)

CAS RN 95-45-4

IUPAC Name: N-[(2Z)-3-nitrosobut-2-en-2-yl]hydroxylamine
Synonyms: dimethylglyoxime
Molecular Weight (g/mol): 116.12
Molecular Formula: C4H8N2O2
InChi Key: OFZZNQXYRRSXOI-ARJAWSKDSA-N
SMILES: C\C(NO)=C(/C)N=O
Quantity 
  • (6)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (4)
Physical Form 
  • (7)
  • (5)
  • (3)
Percent Purity 
  • (3)
  • (7)
Grade 
  • (2)
  • (4)

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (7)
  • (5)
  • (6)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (4)
  • (7)
  • (5)
  • (3)
  • (3)
  • (7)
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.