accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 94838-55-8

CH3H3CCH3OOHNNH2

CAS RN 94838-55-8

IUPAC Name: tert-butyl N-[(4-aminophenyl)methyl]carbamate
Synonyms: tert-butyl N-[(4-aminophenyl)methyl]carbamate
Molecular Weight (g/mol): 222.29
Molecular Formula: C12H18N2O2
InChi Key: UXWQXBSQQHAGMG-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCC1=CC=C(N)C=C1
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.