accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 946387-07-1

H3CCH3NHNCH3H3CSOOClCl

CAS RN 946387-07-1

IUPAC Name: 2,4-dichloro-N-(propan-2-yl)-N-{2-[(propan-2-yl)amino]ethyl}benzene-1-sulfonamide
Synonyms: 2,4-dichloro-N-isopropyl-N-[2-(isopropylamino)ethyl]benzenesulfonamide
Molecular Weight (g/mol): 353.3
Molecular Formula: C14H22Cl2N2O2S
InChi Key: IHYZMEAZAIFMTN-UHFFFAOYSA-N
SMILES: CC(C)NCCN(C(C)C)S(=O)(=O)C1=C(Cl)C=C(Cl)C=C1
Grade 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.