accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 94015-05-1

CAS RN 94015-05-1

IUPAC Name: 4-methylpyridine-3-carboxylate
Synonyms: 4-methylpyridine-3-carboxylate
Molecular Weight (g/mol): 136.13
Molecular Formula: C7H6NO2
InChi Key: ZKUZSTXNVMIDCY-UHFFFAOYSA-M
SMILES: CC1=CC=NC=C1C([O-])=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.