accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 93919-41-6

N(R)NPNHNOPOOP(R)(S)OOOOOOOH2NHOOHONaNaNa

CAS RN 93919-41-6

IUPAC Name: ({[({[5-(2-amino-6-oxo-6,9-dihydro-3H-purin-9-yl)-3-hydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl)oxy](hydroxy)phosphoryl}oxy)phosphonic acid trisodium
Synonyms: deoxyguanosine triphosphate trisodium
Molecular Weight (g/mol): 576.15
Molecular Formula: C10H16N5Na3O13P3
InChi Key: KQAICEQTDXGYDV-UHFFFAOYNA-N
SMILES: [Na].[Na].[Na].NC1=NC(=O)C2=C(N1)N(C=N2)C1CC(O)C(COP(O)(=O)OP(O)(=O)OP(O)(O)=O)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.