accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 936250-20-3

BOOH3CCH3H3CH3CNHNCH3

CAS RN 936250-20-3

IUPAC Name: 5-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Synonyms: 3-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-pyrazole
Molecular Weight (g/mol): 208.07
Molecular Formula: C10H17BN2O2
InChi Key: MSJAEFFWTBMIKT-UHFFFAOYSA-N
SMILES: CC1=C(C=NN1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.