accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 93-08-3

CH3O

CAS RN 93-08-3

IUPAC Name: 1-(naphthalen-2-yl)ethan-1-one
Synonyms: 2-acetonaphthone β-acetonaphthone
Molecular Weight (g/mol): 170.21
Molecular Formula: C12H10O
InChi Key: XSAYZAUNJMRRIR-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C2C=CC=CC2=C1
Boiling Point 
  • (4)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (4)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.