accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 925701-46-8

CH3NOCH3OHNSOONO

CAS RN 925701-46-8

IUPAC Name: 2-(2,6-dimethylmorpholin-4-yl)-N-{5-[6-(morpholin-4-yl)-4-oxo-4H-pyran-2-yl]-9H-thioxanthen-2-yl}acetamide
Synonyms: 2-(2,6-dimethylmorpholin-4-yl)-N-{5-[6-(morpholin-4-yl)-4-oxopyran-2-yl]-9H-thioxanthen-2-yl}acetamide
Molecular Weight (g/mol): 547.67
Molecular Formula: C30H33N3O5S
InChi Key: SCELLOWTHJGVIC-UHFFFAOYNA-N
SMILES: CC1CN(CC(=O)NC2=CC3=C(SC4=C(C=CC=C4C3)C3=CC(=O)C=C(O3)N3CCOCC3)C=C2)CC(C)O1
Grade 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.