CAS RN 924298-51-1
CAS RN 924298-51-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Tocris Bioscience™ ABP 688
High affinity human mGlu5 receptor antagonist
2
3