CAS RN 923032-37-5
CAS RN 923032-37-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
2
Medchem Express Refametinib, MedChemExpress
MedChemExpress Refametinib (BAY 869766; RDEA119) is an orally available, potent, non-ATP-competitive, selective, allosteric MEK1/MEK2 inhibitor with IC50s of 19 nM and 47 nM, respectively.
