CAS RN 922731-01-9
CAS RN 922731-01-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express Omeprazole-d3, MedChemExpress
MedChemExpress Omeprazole-d3 (H 16868-d3) is deuterium labeled Omeprazole. Omeprazole, a proton pump inhibitor (PPI), is available for treatment of acid-related gastrointestinal disorders. Omeprazole shows competitive inhibition of CYP2C19 activity with a Ki of 2 to 6 ÎĽM. Omeprazole also inhibits growth of Gram-positive and Gram-negative bacteria.
