CAS RN 914638-30-5
CAS RN 914638-30-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express IDO-IN-1, MedChemExpress
MedChemExpress IDO-IN-1 is a potent indoleamine 2,3-dioxygenase (IDO) inhibitor with an IC50 of 59 nM.
