CAS RN 891025-25-5
CAS RN 891025-25-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express BAMB-4, MedChemExpress
MedChemExpress BAMB-4(ITPKA-IN-C14) is a new membrane-permeable inhibitor against inositol-1,4,5-trisphosphate-3-kinase A((ITPKA) with IC50 of 37 μM in ADP-Glo Assay.
