CAS RN 887936-68-7
CAS RN 887936-68-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express Temanogrel, MedChemExpress
MedChemExpress Temanogrel is a highly selective 5-HT2A receptor antagonist with a Ki of 4.9 nM.
