accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 88671-89-0

H3CNNNNCl

CAS RN 88671-89-0

IUPAC Name: 2-(4-chlorophenyl)-2-[(1H-1,2,4-triazol-1-yl)methyl]hexanenitrile
Synonyms: myclobutanil
Molecular Weight (g/mol): 288.78
Molecular Formula: C15H17ClN4
InChi Key: HZJKXKUJVSEEFU-UHFFFAOYNA-N
SMILES: CCCCC(CN1C=NC=N1)(C#N)C1=CC=C(Cl)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Color 
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.