accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 880813-36-5

H3CNHNCH3HCl

CAS RN 880813-36-5

IUPAC Name: [(9-ethyl-9H-carbazol-3-yl)methyl](methyl)amine hydrochloride
Synonyms: [(9-ethylcarbazol-3-yl)methyl](methyl)amine hydrochloride
Molecular Weight (g/mol): 274.79
Molecular Formula: C16H19ClN2
InChi Key: IXWVUPURFWFRNE-UHFFFAOYSA-N
SMILES: Cl.CCN1C2=C(C=CC=C2)C2=C1C=CC(CNC)=C2
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.