accessibility menu, dialog, popup

UserName

Nurture Your Progress - Savings to support your research

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 868755-54-8

NNNO

CAS RN 868755-54-8

IUPAC Name: 3-(1H-1,2,4-triazol-1-yl)benzaldehyde
Synonyms: 3-(1,2,4-triazol-1-yl)benzaldehyde
Molecular Weight (g/mol): 173.18
Molecular Formula: C9H7N3O
InChi Key: YGKBHMJLDGOYPI-UHFFFAOYSA-N
SMILES: O=CC1=CC(=CC=C1)N1C=NC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.