accessibility menu, dialog, popup

UserName

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 86-86-2

NH2O

CAS RN 86-86-2

IUPAC Name: 2-(naphthalen-1-yl)acetamide
Synonyms: 1-naphthaleneacetamide
Molecular Weight (g/mol): 185.23
Molecular Formula: C12H11NO
InChi Key: XFNJVKMNNVCYEK-UHFFFAOYSA-N
SMILES: NC(=O)CC1=C2C=CC=CC2=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.