accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 855-97-0

H3COOCH3OOOCH3OH3C

CAS RN 855-97-0

IUPAC Name: 2-(3,4-dimethoxyphenyl)-5,7-dimethoxy-4H-chromen-4-one
Synonyms: 2-(3,4-dimethoxyphenyl)-5,7-dimethoxychromen-4-one
Molecular Weight (g/mol): 342.35
Molecular Formula: C19H18O6
InChi Key: CLXVBVLQKLQNRQ-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=C2C(=O)C=C(OC2=C1)C1=CC=C(OC)C(OC)=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.