accessibility menu, dialog, popup

UserName

Discover Our 3 for 2 Promotion

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 851386-45-3

NFOOHF

CAS RN 851386-45-3

IUPAC Name: 3,6-difluoropyridine-2-carboxylic acid
Synonyms: 3,6-difluoropyridine-2-carboxylic acid
Molecular Weight (g/mol): 159.09
Molecular Formula: C6H3F2NO2
InChi Key: FSXFZWBQCAGFHG-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(F)C=CC(F)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.