accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 85-41-6

OHNO

CAS RN 85-41-6

IUPAC Name: 2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: phthalimide phthalimid o-phthalic imide ftalimmide phenylimide 1,2-benzenedicarboximide 2-diazoindan-1,3-dione benzoimide
Molecular Weight (g/mol): 147.13
Molecular Formula: C8H5NO2
InChi Key: XKJCHHZQLQNZHY-UHFFFAOYSA-N
SMILES: O=C1NC(=O)C2=CC=CC=C12
Boiling Point 
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (4)

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.