CAS RN 83-88-5
CAS RN 83-88-5
Thermo Scientific Chemicals Riboflavin, 98%
CAS: 83-88-5 Molecular Formula: C17H20N4O6 Molecular Weight (g/mol): 376.37 MDL Number: MFCD00005022 InChI Key: AUNGANRZJHBGPY-QTZZOOGMNA-N Synonym: riboflavin,vitamin b2,lactoflavin,riboflavine,vitamin g,lactoflavine,--riboflavin,beflavin,beflavine,flavaxin PubChem CID: 71310809 SMILES: CC1=C(C)C=C2N(C[C@H](O)[C@H](O)[C@H](O)CO)C3=NC(=O)NC(=O)C3=NC2=C1
Thermo Scientific Alfa Aesar Riboflavin, 98%, Thermo Scientific Chemicals
CAS: 83-88-5 Molecular Formula: C17H20N4O6 Molecular Weight (g/mol): 376.37 MDL Number: MFCD00005022 InChI Key: AUNGANRZJHBGPY-QTZZOOGMNA-N Synonym: riboflavin,vitamin b2,lactoflavin,riboflavine,vitamin g,lactoflavine,--riboflavin,beflavin,beflavine,flavaxin PubChem CID: 71310809 IUPAC Name: 7,8-dimethyl-10-[(2S,3S,4R)-2,3,4,5-tetrahydroxypentyl]benzo[g]pteridine-2,4-dione SMILES: CC1=C(C)C=C2N(C[C@H](O)[C@H](O)[C@H](O)CO)C3=NC(=O)NC(=O)C3=NC2=C1
MP Biomedicals Riboflavin USP Grade, MP Biomedicals™
CAS: 83-88-5 Molecular Formula: C17H20N4O6 Molecular Weight (g/mol): 376.37 MDL Number: MFCD00005022 InChI Key: AUNGANRZJHBGPY-QTZZOOGMNA-N Synonym: Vitamin B2,Lactoflavin,Vitaming,7,8-Dimethyl-10- (D-ribo-2,3,4,5- tetrahydroxypentyl) isoalloxazine PubChem CID: 71310809 IUPAC Name: 7,8-dimethyl-10-[(2S,3S,4R)-2,3,4,5-tetrahydroxypentyl]-2H,3H,4H,10H-benzo[g]pteridine-2,4-dione SMILES: CC1=C(C)C=C2N(C[C@H](O)[C@H](O)[C@H](O)CO)C3=NC(=O)NC(=O)C3=NC2=C1
Medchem Express Riboflavin, MedChemExpress
MedChemExpress Riboflavin is an easily absorbed micronutrient with a key role in maintaining health in humans and other animals.

Fisher Bioreagents Riboflavin (Fine, Orange-Yellow Crystals/Electrophoresis), Fisher BioReagents
CAS: 83-88-5 | C17H20N4O6 | 376.37 g/mol