accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 82732-07-8

CAS RN 82732-07-8

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-4-phenylbutanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]-4-phenylbutanoic acid
Molecular Weight (g/mol): 279.34
Molecular Formula: C15H21NO4
InChi Key: MCODLPJUFHPVQP-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CCC1=CC=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.