accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 68547-97-7

HClHOONH

CAS RN 68547-97-7

IUPAC Name: hydrogen 1,2,3,6-tetrahydropyridine-4-carboxylic acid chloride
Synonyms: isoguvacine
Molecular Weight (g/mol): 163.6
Molecular Formula: C6H10ClNO2
InChi Key: SUWREQRNTXCCBL-UHFFFAOYSA-N
SMILES: [H+].[Cl-].OC(=O)C1=CCNCC1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.