accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 68-95-1

CH3ONHOO

CAS RN 68-95-1

IUPAC Name: 1-acetylpyrrolidine-2-carboxylic acid
Synonyms: N-acetylproline
Molecular Weight (g/mol): 157.17
Molecular Formula: C7H11NO3
InChi Key: GNMSLDIYJOSUSW-UHFFFAOYNA-N
SMILES: CC(=O)N1CCCC1C(O)=O
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.