CAS RN 65899-73-2
CAS RN 65899-73-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Thermo Scientific™ Tioconazole, 98+%
Assay: ≥98% CAS: 65899-73-2 Formula Weight: 387.71g/mol Synonym: Thioconazole Format: Powder For Use With (Application): Potent inhibitor of cytochrome-P450 Description: White to Off-white
2
Medchem Express Tioconazole, MedChemExpress
MedChemExpress Tioconazole (UK-20349) is an antifungal imidazole derivative with broad spectrum activity. Tioconazole has inhibitory active aginst several dermatophytes and several yeasts with MIC50s <3.12 mg/L and <9 mg/L, respectively.
