accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 64953-12-4

NaNaH3CONHOOOOHONOOOSNNNNH3C

CAS RN 64953-12-4

IUPAC Name: disodium 7-[2-carboxylato-2-(4-hydroxyphenyl)acetamido]-7-methoxy-3-{[(1-methyl-1H-1,2,3,4-tetrazol-5-yl)sulfanyl]methyl}-8-oxo-5-oxa-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
Synonyms: disodium 7-[2-carboxylato-2-(4-hydroxyphenyl)acetamido]-7-methoxy-3-{[(1-methyl-1,2,3,4-tetrazol-5-yl)sulfanyl]methyl}-8-oxo-5-oxa-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
Molecular Weight (g/mol): 564.44
Molecular Formula: C20H18N6Na2O9S
InChi Key: GRIXGZQULWMCLU-UHFFFAOYNA-L
SMILES: [Na+].[Na+].COC1(NC(=O)C(C([O-])=O)C2=CC=C(O)C=C2)C2OCC(CSC3=NN=NN3C)=C(N2C1=O)C([O-])=O
Quantity 
  • (1)
  • (1)
Color 
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.