accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 64559-06-4

CH3ONH2S

CAS RN 64559-06-4

IUPAC Name: 3-methoxybenzene-1-carbothioamide
Synonyms: 3-methoxybenzenecarbothioamide
Molecular Weight (g/mol): 167.23
Molecular Formula: C8H9NOS
InChi Key: LQSZSWBMLMGWPC-UHFFFAOYSA-N
SMILES: COC1=CC=CC(=C1)C(N)=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.