accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 63155-04-4

CH3ClNH2NH2

CAS RN 63155-04-4

IUPAC Name: 4-chloro-5-methylbenzene-1,2-diamine
Synonyms: 4-chloro-5-methylbenzene-1,2-diamine
Molecular Weight (g/mol): 156.61
Molecular Formula: C7H9ClN2
InChi Key: HOFKXNBVTNUDSH-UHFFFAOYSA-N
SMILES: CC1=CC(N)=C(N)C=C1Cl
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.