accessibility menu, dialog, popup

UserName

Discover Our 3 for 2 Promotion

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 62353-75-7

CH3OOSOH3C

CAS RN 62353-75-7

IUPAC Name: methyl 3-methoxythiophene-2-carboxylate
Synonyms: methyl 3-methoxythiophene-2-carboxylate
Molecular Weight (g/mol): 172.2
Molecular Formula: C7H8O3S
InChi Key: MWADWXJPBMFRGL-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(OC)C=CS1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.