CAS RN 620175-39-5
CAS RN 620175-39-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express AFN-1252, MedChemExpress
MedChemExpress AFN-1252(Debio 1452) is a potent inhibitor of enoyl-acyl carrier protein reductase (FabI), inhibited all clinical isolates of Staphylococcus aureus and Staphylococcus epidermidis at concentrations of ≤0.12 μg/ml.
