accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 6197-30-4

CAS RN 6197-30-4

IUPAC Name: 2-ethylhexyl 2-cyano-3,3-diphenylprop-2-enoate
Synonyms: octocrylene octocrilenum octocrilene octocrileno UV absorber-3 奥克立林 octocrilene (product)
Molecular Weight (g/mol): 361.49
Molecular Formula: C24H27NO2
InChi Key: FMJSMJQBSVNSBF-UHFFFAOYNA-N
SMILES: CCCCC(CC)COC(=O)C(C#N)=C(C1=CC=CC=C1)C1=CC=CC=C1
1x1 image pixel for data collection

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.
1x1 image pixel for data collection