accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 61-76-7

HClH3CNHOHHO

CAS RN 61-76-7

IUPAC Name: hydrogen 3-[1-hydroxy-2-(methylamino)ethyl]phenol chloride
Synonyms: DL-phenylephrine hydrochloride
Molecular Weight (g/mol): 203.67
Molecular Formula: C9H14ClNO2
InChi Key: OCYSGIYOVXAGKQ-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CNCC(O)C1=CC=CC(O)=C1
Percent Purity 
  • (1)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (3)
  • (2)
Grade 
  • (2)

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.