accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 607-01-2

CH3P

CAS RN 607-01-2

IUPAC Name: ethyldiphenylphosphane
Synonyms: phosphine, ethyldiphenyl-
Molecular Weight (g/mol): 214.25
Molecular Formula: C14H15P
InChi Key: WUOIAOOSKMHJOV-UHFFFAOYSA-N
SMILES: CCP(C1=CC=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.