accessibility menu, dialog, popup

UserName

The Ultimate Lab Appliances Promotion

Save Up to 78%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 60079-51-8

ONH2H2NHCl

CAS RN 60079-51-8

IUPAC Name: 2-amino-2-phenylacetamide hydrochloride
Synonyms: acetamide, 2-amino-2-phenyl- hydrochloride
Molecular Weight (g/mol): 186.64
Molecular Formula: C8H11ClN2O
InChi Key: LXOAFHGMXCFTOU-UHFFFAOYNA-N
SMILES: Cl.NC(C(N)=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.