accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 58-46-8

H3CONCH3H3COOH3C

CAS RN 58-46-8

IUPAC Name: 9,10-dimethoxy-3-(2-methylpropyl)-1H,2H,3H,4H,6H,7H,11bH-pyrido[2,1-a]isoquinolin-2-one
Synonyms: tetrabenazine roche brand of tetrabenazine orphan brand of tetrabenazine tetrabenazine product (product) tetrabenazine orphan brand nitoman shire brand of tetrabenazine tetrabenazine (inn) and its salts tetrabenazine product cambridge laboratories brand of tetrabenazine
Molecular Weight (g/mol): 317.43
Molecular Formula: C19H27NO3
InChi Key: MKJIEFSOBYUXJB-UHFFFAOYNA-N
SMILES: COC1=CC2=C(C=C1OC)C1CC(=O)C(CC(C)C)CN1CC2
Physical Form 
  • (2)
  • (2)
Color 
  • (2)
Grade 
  • (2)

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.