accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 56671-28-4

OOOOH

CAS RN 56671-28-4

IUPAC Name: 4-oxo-4,5,6,7-tetrahydro-1-benzofuran-3-carboxylic acid
Synonyms: 4-oxo-6,7-dihydro-5H-1-benzofuran-3-carboxylic acid
Molecular Weight (g/mol): 180.16
Molecular Formula: C9H8O4
InChi Key: FABBWECRHZNMDQ-UHFFFAOYSA-N
SMILES: OC(=O)C1=COC2=C1C(=O)CCC2
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.