CAS RN 547-44-4
CAS RN 547-44-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express Sulfacarbamide, MedChemExpress
MedChemExpress Sulfacarbamide is a blood sugar-lowering drug, also acting on the vegetative nervous system.
